Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031032.2 | NP_112294.1 | 3, 4 | 241 | ||
XM_006251814.2 | XP_006251876.1 | 3, 4 | 902 | |||
GenBank mRNA | BC081778.1 | - | 3, 4 | 241 | 7192655 | |
FQ222145.1 | - | 3, 4 | 303 | |||
Z11558.1 | - | 2, 3 | 161 |