Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202504.1 | NP_001189433.1 | 2 | 146 | ||
NM_001318511.1 | NP_001305440.1 | 2 | 519 | |||
NM_004715.4 | NP_004706.3 | 2 | 519 | |||
NM_048368.3 | NP_430255.2 | 2 | 519 | |||
XM_011526261.1 | XP_011524563.1 | 2 | 497 | |||
GenBank mRNA | AF081287.1 | - | 1 | 67 | ||
AF154115.1 | - | 2 | 455 | |||
BC032515.1 | - | 2 | 173 | |||
BC047999.1 | - | 2 | 519 | |||
BC052576.1 | - | 2 | 111 | |||
BC063447.1 | - | 2 | 474 | 5500032 | ||
DB451780.1 | - | 3 | 270 |