Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itpr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031046.3 | NP_112308.1 | 4 | 577 | ||
GenBank mRNA | AF329470.2 | - | 4 | 552 | ||
AY313846.1 | - | 4 | 331 | |||
X61677.1 | - | 4 | 577 |