Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smad7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030858.1 | NP_110485.1 | 1 | 1956 | ||
XM_006254959.2 | XP_006255021.1 | 1 | 553 | |||
GenBank mRNA | AF042499.1 | - | 1 | 681 | ||
AF159626.1 | - | 1 | 1956 |