Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tp53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030989.3 | NP_112251.2 | 4 | 446 | ||
XM_006246594.2 | XP_006246656.1 | 5 | 875 | |||
XM_006246595.2 | XP_006246657.1 | 4 | 1232 | |||
XM_008767773.1 | XP_008765995.1 | 4 | 453 | |||
GenBank mRNA | AF209191.1 | - | 3 | 338 | ||
BC081788.1 | - | 4 | 375 | 7193583 | ||
BC098663.1 | - | 4 | 404 | 7384467 | ||
CV126174.1 | - | 4 | 446 | |||
FQ232659.1 | - | 4 | 430 | |||
X13058.1 | - | 3 | 338 |