Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alox15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031010.2 | NP_112272.2 | 3 | 381 | ||
XM_006246823.2 | XP_006246885.1 | 5 | 662 | |||
XM_006246824.2 | XP_006246886.1 | 4 | 557 | |||
GenBank mRNA | L06040.1 | - | 3 | 394 | ||
S69383.1 | - | 3 | 381 |