Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031023.1 | NP_112285.1 | 2 | 258 | ||
GenBank mRNA | BC088129.1 | - | 2 | 261 | 7323494 | |
FQ219359.1 | - | 2 | 267 | |||
M95768.1 | - | 2 | 258 |