Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnaq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031036.1 | NP_112298.1 | 3 | 368 | ||
GenBank mRNA | AF234260.1 | - | 3 | 368 | ||
DQ120477.1 | - | 3 | 333 | |||
DQ120478.1 | - | 3 | 333 |