Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gyg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031043.2 | NP_112305.1 | 4 | 504 | ||
GenBank mRNA | AF021343.1 | - | 4 | 507 | ||
BC070944.1 | - | 5 | 545 | 7103004 | ||
FQ215156.1 | - | 2 | 123 | |||
L01793.1 | - | 3 | 324 | |||
U96130.1 | - | 3 | 241 |