Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lifr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031048.1 | NP_112310.1 | 7 | 877 | ||
XM_006232027.2 | XP_006232089.1 | 7 | 1051 | |||
XM_006232029.2 | XP_006232091.1 | 7 | 1712 | |||
XM_008760796.1 | XP_008759018.1 | 7 | 1994 | |||
GenBank mRNA | D86345.1 | - | 7 | 877 |