Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nov | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030868.2 | NP_110495.1 | 5 | 981 | ||
GenBank mRNA | AF171936.1 | - | 5 | 968 | ||
BC072548.1 | - | 5 | 981 | 7105981 |