Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030992.1 | NP_112254.1 | 9 | 1055 | ||
XM_006232205.2 | XP_006232267.1 | 9 | 1830 | |||
XM_006232206.2 | XP_006232268.1 | 10 | 1483 | |||
XM_008760891.1 | XP_008759113.1 | 10 | 1483 | |||
GenBank mRNA | AB003170.1 | - | 9 | 947 | ||
AB003171.1 | - | 9 | 947 | |||
AF017251.1 | - | 8 | 899 | |||
U69550.1 | - | 9 | 1055 | |||
U88986.1 | - | 9 | 967 |