Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031028.3 | NP_112290.2 | 12 | 1546 | ||
XM_006255879.2 | XP_006255941.1 | 12 | 1640 | |||
XM_008772700.1 | XP_008770922.1 | 11 | 1619 | |||
GenBank mRNA | AB016160.1 | - | 7 | 1089 | ||
AB016161.1 | - | 7 | 1089 | |||
AF283276.1 | - | 11 | 1460 | |||
AJ831471.1 | - | 12 | 1544 | |||
AJ831472.1 | - | 12 | 1668 | |||
Y10369.2 | - | 12 | 1518 | |||
Y10370.2 | - | 7 | 1191 |
Set Membership: |
G-protein coupled receptor |