Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnrhr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031038.3 | NP_112300.2 | 1 | 319 | ||
GenBank mRNA | CB775870.1 | - | 1 | 376 | ||
L07646.1 | - | 1 | 305 | |||
X68980.1 | - | 1 | 269 | |||
X76635.1 | - | 1 | 258 |
Set Membership: |
G-protein coupled receptor |