Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253676.1 | NP_001240605.1 | 2 | 699 | ||
NM_004865.3 | NP_004856.1 | 2 | 401 | |||
XM_017011513.1 | XP_016867002.1 | 4 | 974 | |||
GenBank mRNA | AB020881.1 | - | 2 | 316 | ||
AF130312.1 | - | 2 | 344 | |||
AF136570.1 | - | 2 | 348 | |||
AK292320.1 | - | 2 | 654 | |||
AK313645.1 | - | 2 | 369 | |||
BC000381.2 | - | 2 | 369 | 2820491 | ||
BC017559.2 | - | 2 | 312 | 3911352 | ||
BI560657.1 | - | 2 | 406 |