Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031078.2 | NP_112340.1 | 2 | 449 | ||
GenBank mRNA | L41045.1 | - | 2 | 449 |