Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi4kb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031083.1 | NP_112345.1 | 2 | 533 | ||
XM_008761332.1 | XP_008759554.1 | 4 | 875 | |||
XM_008761333.1 | XP_008759555.1 | 3 | 683 | |||
XM_008761334.1 | XP_008759556.1 | 3 | 756 | |||
XM_008761335.1 | XP_008759557.1 | 4 | 875 | |||
XM_008761336.1 | XP_008759558.1 | 2 | 564 | |||
GenBank mRNA | AY724527.1 | - | 2 | 300 | ||
D84667.1 | - | 2 | 533 |