Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Renbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031095.1 | NP_112357.1 | 2 | 166 | ||
XM_006229615.2 | XP_006229677.1 | 3 | 288 | |||
GenBank mRNA | D10233.1 | - | 2 | 166 |