Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031100.1 | NP_112362.1 | 5 | 312 | ||
GenBank mRNA | BC058467.1 | - | 5 | 301 | 6918973 | |
FQ210278.1 | - | 5 | 296 | |||
FQ210394.1 | - | 1 | 14 | |||
FQ211102.1 | - | 1 | 14 | |||
FQ212456.1 | - | 1 | 14 | |||
FQ217196.1 | - | 1 | 14 | |||
FQ217560.1 | - | 1 | 14 | |||
FQ217981.1 | - | 1 | 14 | |||
FQ218098.1 | - | 5 | 296 | |||
FQ220555.1 | - | 5 | 295 | |||
FQ221411.1 | - | 1 | 14 | |||
FQ221714.1 | - | 1 | 14 | |||
FQ221864.1 | - | 5 | 296 | |||
FQ222021.1 | - | 5 | 296 | |||
FQ222201.1 | - | 1 | 14 | |||
FQ222372.1 | - | 1 | 14 | |||
FQ222670.1 | - | 1 | 14 | |||
FQ222847.1 | - | 1 | 14 | |||
FQ223155.1 | - | 1 | 14 | |||
FQ223157.1 | - | 1 | 14 | |||
FQ224407.1 | - | 1 | 14 | |||
FQ224785.1 | - | 1 | 14 | |||
FQ226301.1 | - | 1 | 14 | |||
FQ228167.1 | - | 1 | 14 | |||
FQ229032.1 | - | 5 | 295 | |||
FQ229696.1 | - | 3 | 423 | |||
FQ231791.1 | - | 1 | 14 | |||
FQ233952.1 | - | 5 | 296 | |||
FQ235226.1 | - | 1 | 14 | |||
X87106.1 | - | 5 | 312 |