Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031112.1 | NP_112374.1 | 3 | 136 | ||
XM_006252530.2 | XP_006252592.1 | 3 | 207 | |||
XM_006252531.2 | XP_006252593.1 | 3 | 207 | |||
XM_006252532.2 | XP_006252594.1 | 3 | 207 | |||
XM_006252533.2 | XP_006252595.1 | 3 | 207 | |||
GenBank mRNA | BC058140.1 | - | 3 | 147 | 6919886 | |
FQ221331.1 | - | 3 | 141 | |||
FQ222238.1 | - | 3 | 141 | |||
FQ223039.1 | - | 3 | 141 | |||
FQ225772.1 | - | 3 | 141 | |||
X51538.1 | - | 3 | 136 | |||
X52445.1 | - | 3 | 136 |