Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TESMIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039656.1 | NP_001034745.1 | 6 | 1040 | ||
NM_004923.3 | NP_004914.2 | 6 | 1040 | |||
XM_011545402.1 | XP_011543704.1 | 6 | 1084 | |||
XM_011545403.2 | XP_011543705.1 | 5 | 953 | |||
XM_017018588.1 | XP_016874077.1 | 4 | 886 | |||
GenBank mRNA | AK074817.1 | - | 5 | 910 | ||
AK128308.1 | - | 1 | 1681 | |||
AK292358.1 | - | 6 | 980 | |||
BC027593.1 | - | 6 | 976 | 4838081 | ||
BC064579.1 | - | 6 | 1007 | 5743865 | ||
U86074.1 | - | 5 | 679 |