Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfbr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031132.3 | NP_112394.3 | 2, 3 | 525 | ||
XM_008766690.1 | XP_008764912.1 | 3, 4 | 1081 | |||
GenBank mRNA | GU067683.1 | - | 2, 3 | 274 | ||
L09653.1 | - | 2, 3 | 332 | |||
S67770.1 | - | 2, 3 | 525 |