Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031140.1 | NP_112402.1 | 1 | 568 | ||
GenBank mRNA | BC061847.1 | - | 1 | 608 | 5621065 | |
FQ234991.1 | - | 1 | 322 | |||
X62952.1 | - | 1 | 568 |