Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pax8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031141.2 | NP_112403.2 | 4 | 434 | ||
XM_006233646.2 | XP_006233708.1 | 4 | 1534 | |||
XM_008761610.1 | XP_008759832.1 | 3 | 1870 | |||
XM_008761611.1 | XP_008759833.1 | 3 | 1871 | |||
XM_008761612.1 | XP_008759834.1 | 1 | 1598 | |||
XM_008761613.1 | XP_008759835.1 | 3 | 1866 | |||
GenBank mRNA | BC081779.1 | - | 4 | 434 | 7127710 | |
X94246.1 | - | 3 | 314 |