Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cib1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031145.1 | NP_112407.1 | 4 | 258 | ||
GenBank mRNA | AF136585.1 | - | 4 | 258 | ||
BC091143.1 | - | 4 | 244 | 7308828 |