Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fez1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031066.1 | NP_112328.1 | 2 | 342 | ||
GenBank mRNA | BC087740.1 | - | 2 | 342 | 7314065 | |
U48249.2 | - | 2 | 305 | |||
U63740.1 | - | 2 | 342 |