Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004132.1 | NP_001004132.1 | 6 | 574 | ||
NM_031077.1 | NP_112339.1 | 6 | 933 | |||
XM_006256617.2 | XP_006256679.1 | 6 | 4701 | |||
XM_006256618.2 | XP_006256680.1 | 6 | 934 | |||
XM_006256619.2 | XP_006256681.1 | 6 | 986 | |||
XM_008773037.1 | XP_008771259.1 | 6 | 4704 | |||
GenBank mRNA | BC078711.1 | - | 6 | 574 | 7112874 | |
U36444.1 | - | 6 | 933 |