Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143847.1 | NP_001137319.1 | 4, 5 | 506 | ||
NM_001270604.1 | NP_001257533.1 | 4, 5 | 456 | |||
NM_031079.1 | NP_112341.1 | 3, 4 | 331 | |||
GenBank mRNA | U21101.1 | - | 3, 4 | 331 |