Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163147.1 | NP_001156619.1 | 5 | 736 | ||
NM_001163150.1 | NP_001156622.1 | 2 | 412 | |||
NM_001163152.1 | NP_001156624.1 | 2 | 412 | |||
NM_004956.4 | NP_004947.2 | 6 | 933 | |||
NR_120445.1 | 6 | 628 | ||||
XM_005249635.4 | XP_005249692.1 | 5 | 446 | |||
XM_005249636.4 | XP_005249693.1 | 5 | 369 | |||
XM_011515167.2 | XP_011513469.1 | 4 | 353 | |||
XM_011515168.2 | XP_011513470.1 | 5 | 379 | |||
XM_011515170.2 | XP_011513472.1 | 4 | 231 | |||
GenBank mRNA | AB209202.1 | - | 5 | 383 | ||
AK293802.1 | - | 2 | 100 | |||
AK294572.1 | - | 2 | 310 | |||
AK309190.1 | - | 2 | 148 | |||
AK311089.1 | - | 2 | 179 | |||
AK311194.1 | - | 2 | 144 | |||
AK311707.1 | - | 2 | 310 | |||
AK312863.1 | - | 6 | 615 | |||
AK316007.1 | - | 2 | 339 | |||
BC098403.1 | - | 6 | 607 | 30345383 | ||
BC106762.1 | - | 5 | 426 | 40025690 | ||
BC106763.1 | - | 5 | 425 | 40025693 | ||
CN396908.1 | - | 6 | 560 | |||
CN396911.1 | - | 5 | 365 | |||
CR627389.1 | - | 2 | 167 | |||
DB093597.1 | - | 5 | 378 | |||
DB099964.1 | - | 6 | 489 | |||
DC308837.1 | - | 2 | 412 | |||
DQ204770.1 | - | 88 | ||||
EF632110.1 | - | 324 | ||||
U17163.1 | - | 4 | 194 |
Gene Symbol: TMPRSS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DQ204771.1 | - | 159 |