Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rock1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031098.1 | NP_112360.1 | 3 | 370 | ||
XM_006254400.2 | XP_006254462.1 | 3 | 1122 | |||
XM_006254401.2 | XP_006254463.1 | 3 | 1122 | |||
XM_008771957.1 | XP_008770179.1 | 3 | 1122 | |||
GenBank mRNA | FQ225745.1 | - | 4 | 846 | ||
FQ226663.1 | - | 1 | 76 | |||
U61266.1 | - | 3 | 370 |