Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031099.1 | NP_112361.1 | 3 | 248 | ||
XM_006250585.2 | XP_006250647.1 | 3 | 229 | |||
GenBank mRNA | BC060561.1 | - | 3 | 195 | 6918805 | |
FQ212710.1 | - | 3 | 205 | |||
FQ214281.1 | - | 3 | 196 | |||
FQ214493.1 | - | 3 | 196 | |||
FQ215029.1 | - | 3 | 197 | |||
FQ215486.1 | - | 2 | 69 | |||
FQ219244.1 | - | 3 | 194 | |||
FQ219787.1 | - | 3 | 196 | |||
FQ219848.1 | - | 3 | 196 | |||
FQ220068.1 | - | 3 | 196 | |||
FQ220161.1 | - | 3 | 196 | |||
FQ220170.1 | - | 3 | 196 | |||
FQ220205.1 | - | 3 | 196 | |||
FQ220219.1 | - | 3 | 196 | |||
FQ227439.1 | - | 3 | 196 | |||
FQ228473.1 | - | 3 | 196 | |||
FQ229451.1 | - | 3 | 197 | |||
FQ229816.1 | - | 3 | 194 | |||
FQ229964.1 | - | 3 | 198 | |||
FQ229979.1 | - | 3 | 196 | |||
FQ230017.1 | - | 3 | 194 | |||
FQ230061.1 | - | 3 | 199 | |||
FQ230102.1 | - | 3 | 196 | |||
FQ230128.1 | - | 3 | 224 | |||
FQ230161.1 | - | 3 | 222 | |||
FQ230794.1 | - | 3 | 199 | |||
FQ231729.1 | - | 3 | 194 | |||
FQ232540.1 | - | 3 | 196 | |||
FQ233314.1 | - | 3 | 221 | |||
FQ234171.1 | - | 3 | 226 | |||
M17419.1 | - | 3 | 181 | |||
X06148.1 | - | 3 | 248 |