Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031139.1 | NP_112401.1 | 5, 6 | 1099 | ||
GenBank mRNA | AB035647.1 | - | 5, 6 | 588 | ||
AB035648.1 | - | 5, 6 | 507 | |||
AB035649.1 | - | 4, 5 | 387 | |||
AB035650.1 | - | 4, 5 | 541 | |||
AB035651.1 | - | 2, 3 | 221 | |||
AB047556.1 | - | 5, 6 | 1099 | |||
BC128735.1 | - | 5, 6 | 593 | 7190049 | ||
X90823.1 | - | 3, 4 | 423 |