Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sharpin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031153.2 | NP_112415.1 | 3 | 840 | ||
GenBank mRNA | AF203906.1 | - | 3 | 514 | ||
BC083553.1 | - | 3 | 840 | 7189111 | ||
FQ213170.1 | - | 3 | 458 |