Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270868.1 | NP_001257797.1 | 3 | 453 | ||
NM_001270869.1 | NP_001257798.1 | 2 | 415 | |||
NM_001270870.1 | NP_001257799.1 | 2 | 387 | |||
NM_031073.3 | NP_112335.3 | 3 | 497 | |||
XM_008763294.1 | XP_008761516.1 | 2 | 469 | |||
XM_008763295.1 | XP_008761517.1 | 2 | 449 | |||
GenBank mRNA | BC070504.1 | - | 3 | 497 | 7097740 | |
EV778881.1 | - | 2 | 404 | |||
FM074856.1 | - | 3 | 452 | |||
FM101680.1 | - | 2 | 414 | |||
M34643.1 | - | 2 | 248 |