Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031085.2 | NP_112347.1 | 14 | 2199 | ||
XM_008764720.1 | XP_008762942.1 | 14 | 2081 | |||
GenBank mRNA | BC061763.1 | - | 15 | 2530 | ||
BC081782.1 | - | 14 | 2199 | 7123785 | ||
X68400.1 | - | 14 | 2040 |