Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031139.1 | NP_112401.1 | 8 | 1259 | ||
GenBank mRNA | AB035647.1 | - | 8 | 748 | ||
AB035648.1 | - | 8 | 667 | |||
AB035649.1 | - | 7 | 547 | |||
AB035650.1 | - | 7 | 701 | |||
AB035651.1 | - | 5 | 381 | |||
AB047556.1 | - | 8 | 1259 | |||
BC128735.1 | - | 8 | 753 | 7190049 | ||
X90823.1 | - | 6 | 583 |