Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031140.1 | NP_112402.1 | 3 | 772 | ||
GenBank mRNA | BC061847.1 | - | 3 | 812 | 5621065 | |
FQ220610.1 | - | 1 | 75 | |||
FQ229240.1 | - | 1 | 79 | |||
FQ234991.1 | - | 3 | 526 | |||
X62952.1 | - | 3 | 772 |