Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3gl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031239.2 | NP_112518.1 | 5 | 548 | ||
GenBank mRNA | AB008161.1 | - | 5 | 548 | ||
AF009602.1 | - | 5 | 533 | |||
BC070893.1 | - | 5 | 544 | 7099223 |