Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cds1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031242.2 | NP_112521.2 | 9 | 902 | ||
GenBank mRNA | AB009999.1 | - | 847 | |||
BC127492.1 | - | 9 | 902 | 8373547 | ||
FQ212493.1 | - | 936 |