Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031336.1 | NP_112626.1 | 1 | 722 | ||
GenBank mRNA | AB017820.1 | - | 1 | 722 |