Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031348.2 | NP_112638.2 | 2 | 224 | ||
XM_008761614.1 | XP_008759836.1 | 2 | 247 | |||
GenBank mRNA | AB026057.2 | - | 2 | 135 | ||
BC088128.1 | - | 2 | 224 | 7376326 | ||
FQ227647.1 | - | 2 | 244 | |||
FQ234790.1 | - | 2 | 244 |