Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rara | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031528.2 | NP_113716.2 | 8 | 1466 | ||
XM_008767945.1 | XP_008766167.1 | 8 | 1266 | |||
GenBank mRNA | BC099830.1 | - | 8 | 1466 | 7933868 | |
U15211.1 | - | 8 | 1472 |