Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031561.2 | NP_113749.2 | 360 | |||
GenBank mRNA | AB005743.1 | - | 422 | |||
AF072411.1 | - | 1, 2 | 151 | |||
AF111268.1 | - | 167 | ||||
AF113914.1 | - | 1, 2 | 145 | |||
BC072543.1 | - | 360 | 7105943 | |||
L19658.1 | - | 1, 2 | 200 |
Gene Symbol: LOC103690020 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235962.2 | XP_006236024.1 | 3, 4 | 786 | ||
XM_006235963.2 | XP_006236025.1 | 359 | ||||
XM_006235964.2 | XP_006236026.1 | 3, 4 | 802 | |||
XM_006235965.2 | XP_006236027.1 | 3, 4 | 791 |
Gene Symbol: RGD1565355 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109218.1 | NP_001102688.1 | 151 | |||
GenBank mRNA | BC091104.1 | - | 349 | 7383442 |