Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031568.2 | NP_113756.1 | 3 | 312 | ||
GenBank mRNA | Z67744.1 | - | 3 | 313 |