Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031575.1 | NP_113763.1 | 2 | 122 | ||
GenBank mRNA | D49836.1 | - | 2 | 122 |