Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camkk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031338.1 | NP_112628.1 | 3, 4 | 638 | ||
GenBank mRNA | AB018081.1 | - | 3, 4 | 638 |