Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypla2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031342.1 | NP_112632.1 | 6 | 356 | ||
GenBank mRNA | AB021645.1 | - | 6 | 356 |