Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031346.1 | NP_112636.1 | 3, 4 | 388 | ||
XM_006238206.2 | XP_006238268.1 | 4, 5 | 476 | |||
XM_006238207.2 | XP_006238269.1 | 4, 5 | 468 | |||
XM_006238208.2 | XP_006238270.1 | 3, 4 | 433 | |||
GenBank mRNA | AB023966.1 | - | 3, 4 | 388 |