Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031350.1 | NP_112640.1 | 3, 4 | 384 | ||
GenBank mRNA | AB035306.1 | - | 3, 4 | 384 | ||
BC062046.1 | - | 3, 4 | 475 | 5597999 |