Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vdac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031353.1 | NP_112643.1 | 7 | 665 | ||
XM_006246346.1 | XP_006246408.1 | 6 | 638 | |||
GenBank mRNA | AB039662.1 | - | 7 | 665 | ||
AF048828.1 | - | 7 | 709 | |||
AF268467.1 | - | 6 | 599 | |||
BC060558.1 | - | 7 | 642 | |||
BC072484.1 | - | 7 | 649 | 7105146 | ||
BC087573.1 | - | 7 | 644 | 7304939 | ||
BC087657.1 | - | 7 | 621 | 7312189 | ||
BC104684.1 | - | 7 | 635 | 7462139 | ||
BC127491.1 | - | 7 | 621 | 8370617 |